Hydrocortisone-21-acetate ≥97.0%, micronized USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Hydrocortisone acetate

CO138-1KGBL CO138-100GM CO138-5GM CO138-25GM CO138-5KGBL
95033-516EA 4519 USD
95033-516 95033-518 95033-520 700009-468 95033-514
Hydrocortisone-21-acetate ≥97.0%, micronized USP
Hydrocortisone 21-acetate

Hydrocortisone Acetate, Micronized, USP is a topical corticosteroid product used to help treat eczema, psoriasis and other skin conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Health hazard

Formula: C₂₃H₃₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00037714
CAS Number: 50-03-3

Order Now

Specification Test Results

Assay (Dried Basis) 97.0 - 102.0 %
Specific Rotation @ 20°C +158° to +167°
Loss On Drying max. 1.0 %
Residue On Ignition max. 0.5 %
Organic Impurities To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR