Hydrocodone bitartrate ≥98.0% (CII) USP

H1067-1GM H1067-5GM H1067-25GM H1067-100GMBL
75811-312EA 881.6 USD
75811-312 75811-314 75811-310 75811-308
Hydrocodone bitartrate ≥98.0% (CII) USP
Hydrocodone bitartrate

Hydrocodone Bitartrate (CII), USP is a semi-synthetic opioid derived from codeine and is used as a antitussive and narcotic analgesic via oral administration. It is a DEA CONTROLLED SUBSTANCE - SCHEDULE II drug and does require DEA registration by the customer and completion of DEA purchase notification forms. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₁₈H₂₁NO₃·C₄H₆O₆·2½H₂O
Storage Temperature: Ambient
MDL Number: MFCD00078557
CAS Number: 34195-34-1

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Specific Rotation -79° to -84°
pH of a 2% Solution 3.2 - 3.8
Loss On Drying 7.5 - 12.0 %
Residue On Ignition max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR