Hydrochlorothiazide ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

HY108-25GM
95033-510EA 266.86 USD
95033-510
Hydrochlorothiazide ≥98.0% USP
Hydrochlorothiazide

Hydrochlorothiazide, USP is part of the thiazide class of diuretic drugs used for the treatment of congestive heart failure and hypertension. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₇H₈ClN₃O₄S₂
Storage Temperature: Ambient
MDL Number: MFCD00051765
CAS Number: 58-93-5

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max. 0.5 %
Residue On Ignition max. 0.1 %
Chloride (Cl) max. 0.035 %
Selenium (Se) max. 30 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR