3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture)

Synonyms: CoppertoneSalicylic Acid 3,3,5-Trimethylcyclohexyl EsterHomosalate2-Hydroxybenzoic Acid 3,3,5-Trimethylcyclohexyl Ester3,3,5-Trimethylcyclohexyl 2-Hydroxybenzoate3,3,5-Trimethylcyclohexyl 2-hydroxybenzoate

H1187-200KGB H1187-100GM H1187-200KGB H1187-100GM H1187-25GM H1187-25GM
700011-778EA 185.84 USD
700011-778 CA700011-776 700011-774 700011-776 CA700011-778 CA700011-774
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture)
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture)

Homosalate is an organic compound most often used in sunscreens as a chemical UV filter.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₂₂O₃
MW: 262.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019377
CAS Number: 118-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR