Guaiacwood oil

Supplier: SPECTRUM CHEMICAL MFG CORP

G1183-100ML
700010-632EA 149 USD
700010-632
Guaiacwood oil
Guaiacwood oil

Guaiacwood Oil is an essential oil commonly used as a fragrance ingredient for soap and fine fragrances.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD01772822
CAS Number: 8016-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR