(+)-Griseofulvin ≥97.0%, micronized Ph. Eur., BP

Synonyms: Griseofulvin

G1394-100GM G1394-500GM G1394-25GM G1394-5KGBL G1394-1KGBL
75811-256EA 3179.84 USD
75811-256 75811-258 75811-254 75811-260 75811-262
(+)-Griseofulvin ≥97.0%, micronized Ph. Eur., BP
(+)-Griseofulvin

Griseofulvin, Micronized, BP, EP is an antifungal drug used most often to treat skin infections such as jock itch, athletes foot, ringworm and fungal infections of the scalp, toenails, and fingernails. The BP and EP grades indicate it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical BP and EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. (CAS# 126-07-8, Molecular Formula: C17H17ClO6) Griseofulvin, Micronized, BP, EP


GHS hazard : Exclamation mark;Health hazard

Formula: C₁₇H₁₇ClO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00082343
CAS Number: 126-07-8

Order Now

Specification Test Results

Assay (Dried Basis) 97.0 - 102.0%
Appearance of Solution To Pass Test
Specific Optical Rotation +354° to +364°
Acidity To Pass Test
Related Substances To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR