Glycofural

Synonyms: GlycofurolTetrahydrofurfuryl alcohol polyethylene glycol ether

G2001-25KGBL G2001-25GM
700012-516EA 15321.51 USD
700012-516 700012-514
Glycofural
Glycofural

Glycofurol is used as solvent to dissolve water-insoluble compounds.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₁₄O₃
Density: 1.069
Storage Temperature: Ambient
MDL Number: MFCD00801146
CAS Number: 31692-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR