Glyceryl tribenzoate ≥95%

G1476-250GM
700005-576EA 176.69 USD
700005-576
Glyceryl tribenzoate ≥95%
Glyceryl tribenzoate
Storage Temperature: Ambient
MDL Number: MFCD00020677
CAS Number: 614-33-5

Order Now

Specification Test Results

Assay Approx. 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR