Glucosamine sulfate, stabilized with sodium chloride stabilized
Stabilization: Stabilised with potassium chloride
G1125-25KGBL
700014-368EA 2319.09 USD
700014-368
Glucosamine sulfate, stabilized with sodium chloride stabilized
Glucosamine sulfate, stabilized with sodium chloride
Glucosamine Sulfate, Stabilized with Potassium Chloride is an amino sugar and plays an important role in the biochemical synthesis of glycosylated lipids and proteins. It is most commonly used as a non-vitamin dietary supplement.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.