Glipizide ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: N-[4-(3-Cyclohexylureidosulfonyl)phenethyl]-5-methyl-2-pyrazinecarboxamideN-(4-(N-(Cyclohexylcarbamoyl)sulphamoyl)phenethyl)-5-methylpyrazine-2-carboxamide1-Cyclohexyl-3-{4-[2-(5-methylpyrazine-2-carboxamido)ethyl]phenylsulfonyl}ureaN-[4-(3-Cyclohexylureidosulphonyl)phenethyl]-5-methyl-2-pyrazinecarboxamide1-Cyclohexyl-3-{4-[2-(5-methylpyrazine-2-carboxamido)ethyl]phenylsulphonyl}ureaN-(4-(N-(Cyclohexylcarbamoyl)sulfamoyl)phenethyl)-5-methylpyrazine-2-carboxamide

G1120-1KGBL G1120-5GM
95033-382EA 696 USD
95033-382 95033-378
Glipizide ≥98.0% USP
Glipizide

Glipizide, USP is an oral diabetes medicine that helps control blood sugar levels by helping your pancreas produce insulin. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₂₁H₂₇N₅O₄S
MW: 445.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00072159
CAS Number: 29094-61-9

Order Now

Specification Test Results

Assay (Dried Basis) 98.0- 102.0 %
Loss On Drying max. 1.0 %
Residue On Ignition max. 0.4 %
Heavy Metals max. 0.005 %
Related Compounds To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR