Ghatti gum

Supplier: SPECTRUM CHEMICAL MFG CORP

GH105-125GM GH105-500GM
700000-056EA 85.42 USD
700000-056 700000-058
Ghatti gum
Ghatti gum

Ghatti Gum, from the Anogeissus latifolia species of trees, is used in India for tanning and for calico printing; it is also known as Indian gum.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00131248
CAS Number: 9000-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR