Galaxolide(1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran) solution 50% in diethyl phthalate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4,6,6,7,8,8-Hexamethyl-1,3,4,6,7,8-hexahydrocyclopenta[g]isochromeneHexamethylindanopyranAbbalide

G1438-100GM
700009-124EA 122 USD
700009-124
Galaxolide(1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran) solution 50% in diethyl phthalate
Galaxolide(1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran)

Galaxolide, 50 Percent Solution in Diethyl Phthalate, also known as Hexahydrohexamethyl cyclopentabenzopyran, is a synthetic ingredient used in fragrances and is known for its woody scent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₈H₂₆O
MW: 258.41 g/mol
Density: 1.044
Storage Temperature: Ambient
MDL Number: MFCD00217003
CAS Number: 1222-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR