Furosemide ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Chloro-N-furfuryl-5-sulphamoylanthranilic acid5-(Aminosulphonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid5-(Aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid

F1133-1KGBL F1133-5GM F1133-100GM F1133-500GM F1133-25GM
95033-340EA 1147 USD
95033-340 95033-342 95033-344 95033-346 95033-348
Furosemide ≥98.0% USP
Furosemide

Furosemide, USP is used to reduce extra fluid in the body (edema) caused by conditions such as heart failure, liver disease, and kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Health hazard

Formula: C₁₂H₁₁ClN₂O₅S
MW: 330.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010549
CAS Number: 54-31-9

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max. 1.0 %
Residue On Ignition max. 0.1 %
Heavy Metals max. 20 ppm
Organic Impurities To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR