Ethylenediaminetetraacetic acid diammonium salt hydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: EDTA diammonium salt hydrate

D2304-25GM D2304-100GM
700007-262EA 57.77 USD
700007-262 700007-260
Ethylenediaminetetraacetic acid diammonium salt hydrate
Ethylenediaminetetraacetic acid diammonium salt hydrate

Ethylenediaminetetraacetic Acid Diammonium Salt is typically known as EDTA and is an aminopolycarboxylic acid. It is a colorless solid and soluble in water. It is usually used to dissolve limescale.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₂₂N₄O₈·xH₂O
MW: 326.31 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150463
CAS Number: 20824-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR