Estrone ≥97.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

ES200-1KGBL ES200-5GM ES200-25GM
95033-140EA 361.01 USD
95033-140 95033-136 95033-138
Estrone ≥97.0% USP
Estrone

Estrone, USP is a sex hormone that is used in hormone replacement therapy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Health hazard

Formula: C₁₈H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00003620
CAS Number: 53-16-7

Order Now

Specification Test Results

Assay (Dried Basis) 97.0 - 103.0 %
Clarity of Solution To Pass Test
Specific Rotation [A]D +158 to +165
Loss On Drying max. 0.5 %
Equilenin and Equilin To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR