L-Adrenaline ≥97.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: L-Epinephrine

EP130-1KGBL
95031-102EA 15027 USD
95031-102
L-Adrenaline ≥97.0% USP
L-Adrenaline

Epinephrine, USP is also known as adrenaline and is a hormone and neurotransmitter. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Skull and crossbones;Exclamation mark
UN2811

Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002204
CAS Number: 51-43-4
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay (Dried Basis) 97.0 - 100.5%
Specific Rotation [A]D -50.0° to -54.0
Loss On Drying max. 2.0%
Residue On Ignition Negligible
Adrenalone To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR