Econazole nitrate ≥99.0%

Synonyms: 1-[2-(4-Chlorophenylmethoxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole nitrateEcostatin1-[2-(4-Chlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole Nitrate

E1107-40KGBL E1107-100GM E1107-5GM E1107-1KG E1107-25GM
95033-040EA 399.65 USD
95033-040 95033-042 700008-612 95033-036 95033-038
Econazole nitrate ≥99.0%
Econazole nitrate

Econazole Nitrate, BP is an antifungal medication. All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₁₈H₁₅Cl₃N₂O·HNO₃
MW: 444.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058160
CAS Number: 68797-31-9

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 101.0%
Related Substances To Pass Test
Loss On Drying max. 0.5%
Sulphated Ash max. 0.1%
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR