D-Saccharic acid potassium salt ≥98.5%
Supplier: SPECTRUM CHEMICAL MFG CORP
S1468-25GM
S1468-100GM
700007-868EA
362.98
USD
700007-868
700007-866
D-Saccharic acid potassium salt ≥98.5%
D-Saccharic acid potassium salt
D-Saccharic Acid Monopotassium Salt, also known as glucaric acid, is derived by oxidizing a sugar with nitric acid. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
C₆H₉KO₈ Storage Temperature: Ambient |
CAS Number:
576-42-1 |
Specification Test Results
Assay | 98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...