Dowtherm® SR-1
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: OctadeceneEthylene Glycol with Corrosion inhibitors
D1264-20LT
71001-990EA 1004 USD
71001-990
Dowtherm® SR-1
Dowtherm® SR-1
Dowtherm(R) SR-1 is also known as octadecene. It is an alkene and a long chain hydrocarbon. It has been used in the synthesis of colloidal quantum dots. It is considered an inexpensive solvent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.