DL-Tartaric acid ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Racemic Acid
T2808-500GM
T2808-25GM
T2808-100GM
700006-124EA
178
USD
700006-124
700006-122
700006-120
DL-Tartaric acid ≥99%
DL-Tartaric acid
DL-Tartaric Acid, also known as 2,3-dihydroxysuccinic acid, is added to other foods to give a sour taste, and is used as an antioxidant..
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₄H₆O₆ MW: 150.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00071626 CAS Number: 133-37-9 |
Specification Test Results
Assay | 99 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...