Dioctadecyl-3,3'-thiodipropionate ≥90%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dioctadecyl 3,3'-thiodipropanoateDistearyl 3,3'-Thiodipropionate3,3'-Thiodipropionic Acid Dioctadecyl Ester

D3833-100GM
700014-170EA 113 USD
700014-170
Dioctadecyl-3,3'-thiodipropionate ≥90%
Dioctadecyl-3,3'-thiodipropionate
Formula: C₄₂H₈₂O₄S
MW: 683.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026684
CAS Number: 693-36-7

Order Now

Specification Test Results

Assay Approx. 90 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR