Diphenhydramine hydrochloride ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-(Benzhydryloxy)-N,N-dimethylethylamine Hydrochloride2-Diphenylmethoxy-N,N-dimethylethylamine Hydrochloride2-(Benzhydryloxy)-N,N-dimethylethanamine hydrochloride

DI160-50KGBL DI160-12KGBL DI160-5GM DI160-500GM DI160-100GM
700009-850EA 1703.68 USD
700009-850 95032-950 700009-852 95032-954 95032-952
Diphenhydramine hydrochloride ≥98.0% USP
Diphenhydramine hydrochloride

Diphenhydramine Hydrochloride, USP is an antihistamine that includes sedative attributes to treat allergies. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₁₇H₂₁NO
MW: 291.82 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012479
CAS Number: 147-24-0

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Identification To Pass Test
Acidity or Alkalinity To Pass Test
Loss On Drying max. 0.5 %
Residue On Ignition max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR