Dimenhydrinate USP

Synonyms: Amosyt8-Chloro-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione 2-(benzhydryloxy)-N,N-dimethylethanamine salt

DI127-500GMB DI127-100GM
95032-938EA 832.62 USD
95032-938 95032-942
Dimenhydrinate USP
Dimenhydrinate

Dimenhydrinate, USP is used to prevent and treat nausea, vomiting, and dizziness caused by motion sickness. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₁₇H₂₁NO·C₇H₇ClN₄O₂
MW: 469.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054265
CAS Number: 523-87-5

Order Now

Specification Test Results

Assay (C₁₇H₂₁NO; Dried Basis) 53.0 - 55.5 %
Assay (C₇H₇ClN₄O₂; Dried Basis) 44.0 - 47.0 %
Melting Range 102 - 107 °C
Loss On Drying max. 0.5 %
Residue On Ignition max. 0.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR