Dihydroergotamine mesylate ≥97.0% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Dihydroergotamine methanesulfonate
, Dihydroergotamine methanesulphonate
D1165-100GMB
D1165-5GM
D1165-1GM
D1165-250MG
75810-986EA
2107
USD
75810-986
75810-984
75810-982
95032-920
Dihydroergotamine mesylate ≥97.0% USP
Dihydroergotamine mesylate
Dihydroergotamine Mesylate, USP is an ergot alkaloid medication used to treat active migraine headaches. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Health hazard
Specification Test Results
Assay (Dried Basis) | 97.0 - 103.0% |
Specific Rotation | -16.7° to -22.7 |
pH of a 0.1% Solution @ 20°C | 4.4 - 5.4 |
Loss On Drying | max. 4.0% |
Related Alkaloids | max. 2.0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...