Diclofenac sodium salt ≥99.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: [o-(2,6-dichloroanilino) phenyl]acetic acid sodium saltSodium [o-[(2,6-dichlorophenyl) amino] phenyl] acetate2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium saltGP-458402-[(2,6-Dichlorophenyl)amino]-benzeneacetic acid monosodium salt

D1287-500GM D1287-100GM D1287-25GM
95031-062EA 390 USD
95031-062 95031-064 95031-060
Diclofenac sodium salt ≥99.0% USP
Diclofenac sodium salt

Diclofenac Sodium, USP is a nonsteroidal anti-inflammatory drug used to reduce inflammation and is as an analgesic. All Spectrum Chemical USP products are manufactured, packa ged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Skull and crossbones;Health hazard
UN2811

Formula: C₁₄H₁₀Cl₂NNaO₂
MW: 318.1 g/mol
Storage Temperature: Ambient
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 101.0%
Color of Solution To Pass Test
Clarity of Solution To Pass Test
pH of a 1 in 100 Solution 7.0 - 8.5
Chromatographic Purity To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR