Dibutyl adipate ≥96%

Synonyms: Adipic Acid Dibutyl Ester

D1467-250ML
700005-804EA 65.57 USD
700005-804
Dibutyl adipate ≥96%
Dibutyl adipate

GHS hazard : Exclamation mark

Formula: C₁₄H₂₆O₄
MW: 258.36 g/mol
Density: 0,962
Storage Temperature: Ambient
MDL Number: MFCD00009451
CAS Number: 105-99-7

Order Now

Specification Test Results

Assay Approx. 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR