Disodium hydrogen phosphate dodecahydrate ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dibasic sodium phosphate dodecahydrateSodium phosphate dibasic dodecahydrateSodium hydrogen phosphate dodecahydratedi-Sodium hydrogen orthophosphate dodecahydrateSodium monohydrogen phosphate dodecahydrate

S2056-50KGBL
700001-668EA 4519 USD
700001-668
Disodium hydrogen phosphate dodecahydrate ≥98.0% USP
di-Sodium hydrogen phosphate dodecahydrate

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₂HPO₄·12H₂O
MW: 358.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149181
CAS Number: 10039-32-4

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 100.5 %
Loss On Drying 55.0 - 64.0 %
Insoluble Substances max. 0.4 %
Chloride (Cl) max. 0.06 %
Sulfate max. 0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR