Diazolidinyl urea ≥95%

Synonyms: 1-(1,3-Bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)-1,3-bis(hydroxymethyl)urea1,3-Bis(hydroxymethyl)-5-[1,3-bis(hydroxymethyl)ureido]hydantoin

D2841-25GM D2841-250GM D2841-20KGBL
700008-608EA 205.26 USD
700008-608 700008-606 700008-610
Diazolidinyl urea ≥95%
Diazolidinyl urea

Diazolidinyl Urea is an antimicrobial preservative most often used in cosmetics, skin care products, shampoos and conditioners, as well as a wide range of products including bubble baths, baby wipes and household detergents.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₁₄N₄O₇
MW: 278.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03547942
CAS Number: 78491-02-8

Order Now

Specification Test Results

Assay Approx. 95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR