Diacetin

Synonyms: Glycerol alpha,alpha'-Diacetate2-Hydroxypropane-1,3-diyl diacetate

D3159-500GM D3159-100GM D3159-5GM
700004-460EA 89.37 USD
700004-460 700004-464 700004-462
Diacetin
Diacetin

Caution: Storage temperature - HYGROSCOPIC: Keep tightly closed.

Formula: C₇H₁₂O₅
MW: 176.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008717
CAS Number: 25395-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR