D-Glucoheptono-1,4-lactone ≥99%

G3399-100GM G3399-25GM
700008-392EA 100.2 USD
700008-392 700008-390
D-Glucoheptono-1,4-lactone ≥99%
D-Glucoheptono-1,4-lactone

D-Glucoheptono-1,4-lactone is used as a cosmetic ingredient for skin conditioning.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00005390
CAS Number: 60046-25-5

Order Now

Specification Test Results

Assay 99% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR