DNA (Deoxyribonucleic acid)

DE115-100GM DE115-25GM DE115-1KGBL
700014-186EA 166.09 USD
700014-186 700014-184 700014-182
DNA (Deoxyribonucleic acid)
DNA (Deoxyribonucleic acid)

Deoxyribonucleic Acid is a nucleic acid composed of three macromolecules important for all forms of life. The molecule encodes the genetic map used in functioning living viruses and organisms.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00130922
CAS Number: 68938-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR