Denatonium saccharide ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

D1178-1GM D1178-25GM D1178-5GM
700014-028EA 268 USD
700014-028 700014-026 700014-024
Denatonium saccharide ≥95%
Denatonium saccharide

Denatonium Saccharide, also known as denatonium benzoate, is used to denature ethanol and used in rubbing alcohol as an inactive ingredient.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₈H₃₃N₃O₄S
MW: 507.6443 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00214097
CAS Number: 90823-38-4

Order Now

Specification Test Results

Assay Approx. 95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR