D-δ-Tocopherol

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2R)-2,8-dimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol(+)-δ-Tocopherol

T1387-1GM
700009-390EA 170 USD
700009-390
D-δ-Tocopherol
D-delta-Tocopherol

D-delta-Tocopherol is one of the naturally occurring forms of Vitamin E.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₇H₄₆O₂
MW: 402.663 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20486794
CAS Number: 119-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR