Danazol ≥97.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

D1103-5GM D1103-1KGBL D1103-25GM D1103-500GMBL D1103-250MG D1103-1GM
95032-792EA 5500 USD
95032-792 95032-790 95032-786 95032-784 95032-794 75810-980
Danazol ≥97.0% USP
Danazol

Danazol, USP is used in women to treat pelvic pain and infertility. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Health hazard

Formula: C₂₂H₂₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00056838
CAS Number: 17230-88-5

Order Now

Specification Test Results

Assay (Dried Basis) 97.0 - 102.0 %
Assay (Dried Basis) 97.0 - 102.0 %
Specific Rotation +21 to +27
Specific Rotation +21 to +27
Chromatographic Purity To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR