α-D-Galacturonic acid hydrate ≥80%

Synonyms: (2S,3R,4S,5R)-2,3,4,5-Tetrahydroxy-6-oxohexanoic acid hydrate

G3072-5GM
700008-498EA 77.64 USD
700008-498
α-D-Galacturonic acid hydrate ≥80%
alpha-D-Galacturonic acid hydrate

D-(+)-Galacturonic Acid, Monohydrate, also known simply as D-Galacturonic acid, is an oxidized form of D-galactose and a sugar acid. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₆H₁₀O₇·xH₂O
MW: 194.14 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00006618
CAS Number: 91510-62-2

Order Now

Specification Test Results

Assay Approx. 80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR