Quinizarin green ≥96%

Synonyms: Solvent Green 3D and C Green No. 61,4-Bis(p-tolylamino)anthracene-9,10-dione1,4-Bis(p-toluidino)-9,10-anthraquinone

D1126-25GM D1126-100GM D1126-500GM
700010-260EA 255.76 USD
700010-260 700010-264 700010-262
Quinizarin green ≥96%
1,4-Bis(p-toluidino)-9,10-anthraquinone

D and C Green No. 6 | Non-graded D and C Green No. 6 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₈H₂₂N₂O₂
MW: 418.496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001195
CAS Number: 128-80-3

Order Now

Specification Test Results

Assay 96 % min.
Volatile Matter (@ 135°C) max. 2.0 %
Water Soluble Matter max. 0.3 %
Matter Insoluble in Carbon[Ntetrachloride max. 1.5 %
Dihydroxyanthraquinone max. 0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR