Cyclohexylamine ≥99.0%

Synonyms: Aminocyclohexane

C2236-2.5LTPV C2236-500ML
10841-764EA 252.77 USD
CA11027-320 10841-764
Cyclohexylamine ≥99.0%
Cyclohexylamine

Cyclohexylamine, also known as aminocyclohexane, is used as an intermediate in synthesis of other organic compounds. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


GHS hazard : Corrosion;Flame;Skull and crossbones;Exclamation mark;Health hazard
UN2357

Formula: C₆H₁₃N
MW: 99.18 g/mol
Density: 0.867
Storage Temperature: Ambient
MDL Number: MFCD00001486
CAS Number: 108-91-8
UN: 2357
ADR: 8.3,II

Order Now

Specification Test Results

Assay 99.0 %
Appearance Colorless Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR