SPCMC1470
C1470-5GM C1470-1GM
10841-760EA 83.27 USD
10841-760 10841-762
SPCMC1470
Cycloheximide, also known as hizarocin, is used in biomedical research. It inhibits protein synthesis in eukaryotic cells studied in vitro. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
GHS hazard : Skull and crossbones;Exclamation mark;Health hazard
UN2811