Cyclobenzaprine hydrochloride ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 3-(5H-Dibenzo[a,d][7]annulen-5-ylidene)-N,N-dimethylpropan-1-amine hydrochloride

C1570-5GMGL C1570-500GMB C1570-100GMGL C1570-25GMGL
75810-836EA 6216 USD
75810-836 75810-840 95032-764 75810-838
Cyclobenzaprine hydrochloride ≥98.0% USP
Cyclobenzaprine hydrochloride

Cyclobenzaprine Hydrochloride, USP is a muscle relaxant used to treat skeletal muscle conditions such as pain or injury. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Skull and crossbones;Exclamation mark
UN2811

Formula: C₂₀H₂₁N·HCl
MW: 311.8484 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079039
CAS Number: 6202-23-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max.1.0 %
Residue On Ignition max. 0.1 %
Heavy Metals max. 0.001 %
Organic Impurities To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR