Croscarmellose sodium NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Croscarmellose Sodium Modified Cellulose GumCarmellose SodiumSodium carboxymethyl cellulose

C1366-2.5KG C1366-25KGBL C1366-500GM
95032-728EA 826 USD
95032-728 95032-730 700013-728
Croscarmellose sodium NF
Croscarmellose sodium

Croscarmellose Sodium, NF is used commonly as a FDA-approved disintegrant in pharmaceutical manufacturing. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. These are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Storage temperature - Keep tightly closed

Formula: C₂₈H₃₀Na₈O₂₇
MW: 982.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00217760
CAS Number: 74811-65-7

Order Now

Specification Test Results

pH of a 1 % Dispersion 5.0 - 7.0
Degree of Substitution (Dried Basis) 0.60 - 0.85
Residue On Ignition 14.0 - 28.0 %
Settling Volume To Pass Test
Loss On Drying max. 10.0 %
Sodium Chloride and Sodium Glycolate 0.5%
Content of Water-Soluble Material 10%
Microbial Limits To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR