Copper(II) gluconate ≥98.0% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Copper(II) D-gluconate
, Copper Gluconate
, Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
, Gluconic Acid Copper(II) Salt
Copper Gluconate, FCC is used as a nutrient and dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₂H₂₂CuO₁₄ MW: 453.84 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00075297 CAS Number: 527-09-3 |
Specification Test Results
Assay (C₁₂H₂₂O₁₄Cu) | 98.0- 102.0 % |
Lead (Pb) | max. 5 mg/kg |
Reducing Substances | max. 1.0 % |
Identification | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...