Copaiba balsam

CO132-500GM CO132-125GM
700013-970EA 120.73 USD
700013-970 700013-972
Copaiba balsam
Copaiba balsam

Copaiba Balsam, a stimulant oleoresin obtained from South American leguminous trees, is also known as turpentine and can be used in making lacquers and varnishes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00146552
CAS Number: 8001-61-4

Order Now

Specification Test Results

Acid Number 28 - 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR