Cocoa butter, deodorized

C1657-25KGBL
700013-834EA 799.99 USD
700013-834
Cocoa butter, deodorized
Cocoa butter

Cocoa Butter, Deodorized, a butter like substance that is obtained from cocoa beans. It is a vegetable fat that is edible and highly prized for its cocoa flavor and aroma, especially in the chocolate industry. It is used in nearly every chocolate based food but is nowadays also used for its physical properties in pharmaceutical formulations. It is also used in cosmetic products and skin care creams aimed at moisturizing the skin.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00146492
CAS Number: 8002-31-1

Order Now

Specification Test Results

Appearance Yellowish White
Iodine Value 33 - 42
Saponification Value 188 - 198
Free Fatty Acids max. 0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR