Citalopram hydrobromide ≥98.0% USP
|
|
Synonyms:
1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Hydrobromide
C2408-1KGBL
C2408-100GMBL
75810-878EA
7502.2
USD
75810-878
75810-876
Citalopram hydrobromide ≥98.0% USP
Citalopram hydrobromide
Citalopram Hydrobromide, USP is used as an antidepressant drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Exclamation mark;Health hazard
Formula:
C₂₀H₂₁FN₂O MW: 405.31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02101306 CAS Number: 59729-32-7 |
Specification Test Results
Assay (Anhydrous Basis) | 98.0 - 102.0% |
Completeness of Solution | To Pass Test |
Specific Rotation | -0.2° to +0.2° |
pH | 5.5 - 6.5 |
Water Determination | max. 0.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...