cis-Stilbene ≥93%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Isostilbene(Z)-1,2-Diphenylethenecis-1,2-Diphenylethylenecis-1,2-Diphenylethene

S2746-5GM S2746-25GM S2746-1GM
10860-336EA 289 USD
10860-336 10860-332 10860-334
cis-Stilbene ≥93%
cis-Stilbene
Formula: (C₆H₅)CHCH(C₆H₅)
MW: 180.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004788
CAS Number: 645-49-8

Order Now

Specification Test Results

Purity (GC) 93 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR