Ciclopirox olamine ≥97.5% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
6-Cyclohexyl-1-hydroxy-4-methylpyridin-2(1H)-one 2-aminoethanol salt
C1486-1GM
C1486-5GM
C1486-25GM
C1486-500GMB
C1486-100GM
75810-824EA
125
USD
75810-824
75810-822
700009-104
700009-106
700009-108
Ciclopirox olamine ≥97.5% USP
Ciclopirox olamine
Ciclopirox Olamine, USP is a synthetic antifungal agent for topical dermatologic treatment of superficial mycoses. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
GHS hazard : Exclamation mark
Formula:
C₁₂H₁₇NO₂·C₂H₇NO MW: 268.3520 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078997 CAS Number: 41621-49-2 |
Specification Test Results
Assay | 97.5 - 101.5% |
pH of a 1% Solution | 8.0 - 9.0 |
Monoethanolamine | 223 - 230 MG/G |
Residue On Ignition | max. 0.1% |
Heavy Metals | max. 0.001% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...