Chlorpheniramine maleate ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Chlorphenamine maleate

CH133-50KGBL CH133-1KG CH133-100GM CH133-25GM
700009-808EA 19940 USD
700009-808 95032-544 95032-542 95032-546
Chlorpheniramine maleate ≥98.0% USP
Chlorpheniramine maleate

Chlorpheniramine Maleate, USP is a first generation alkylamine antihistamine that is prevents the appearance of allergic conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: C₁₆H₁₉ClN₂
MW: 390.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069225
CAS Number: 113-92-8

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Specific Rotation, [A]D25 -0.10 to +0.10
Loss On Drying max. 0.5 %
Residue On Ignition max. 0.2 %
Organic Impurities To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR