4-Chloro-3,5-xylenol ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Chloroxylenol4-Chloro-3,5-dimethylphenol2-Chloro-5-hydroxy-3-xylene

CH136-5GM CH136-500GM CH136-25KGBL CH136-100GM CH136-25GM CH136-50KGBL CH136-12KGBL
95032-538EA 499 USD
95032-538 95032-540 75810-938 95032-536 75810-936 95032-534 700003-356
4-Chloro-3,5-xylenol ≥98.5% USP
4-Chloro-3,5-xylenol

Chloroxylenol, USP is a broad spectrum antimicrobial chemical compound used to control bacteria, algae, fungi and virus. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: C₈H₉ClO
MW: 156.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002324
CAS Number: 88-04-0

Order Now

Specification Test Results

Assay 98.5 % min.
Water max.0.5 %
Tetrachloroethylene max. 0.4 %
Residue On Ignition max. 0.1 %
Iron max. 0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR