Chlorhexidine dihydrochloride ≥98.0%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,1'-Hexamethylenebis[5-(4-chlorophenyl)biguanide] DihydrochlorideN1,N14-Bis(4-chlorophenyl)-3,12-diimino-2,4,11,13-Tetraazatetradecanediimidamide dihydrochloride

C2269-5GM C2269-25GM C2269-1GM
700007-892EA 80.89 USD
700007-892 700007-886 700007-888
Chlorhexidine dihydrochloride ≥98.0%
Chlorhexidine dihydrochloride

Chlorhexidine Dihydrochloride is an odorless, white crystalline powder that is a synthetic preservative and antiseptic that functions as a cosmetic biocide and oral care agent. It is used in the formulation of eye makeup, makeup foundations, mouthwashes, hair dyes and bleaches, and other skin care and hair care products

Formula: C₂₂H₃₀Cl₂N₁₀
MW: 578.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00068998
CAS Number: 3697-42-5

Order Now

Specification Test Results

Purity (HPLC; Area %) 98.0 % min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR