Ceresin wax

Supplier: SPECTRUM CHEMICAL MFG CORP

CE116-2.5KG CE116-500GM CE116-50LBBL CE116-12KG
700013-962EA 1088 USD
700013-962 700013-964 700013-966 700013-968
Ceresin wax
Ceresin wax

Ceresin Wax, also known as ceresin or cerin, is a wax that comes from ozokerite through a purifying process.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
CAS Number: 8001-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR